Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301779.1 | NP_001288708.1 | 8 | 1471 | ||
NM_001301780.1 | NP_001288709.1 | 7 | 1327 | |||
NM_001301819.1 | NP_001288748.1 | 6 | 1188 | |||
NM_003455.3 | NP_003446.2 | 9 | 1523 | |||
XM_005271659.1 | XP_005271716.1 | 9 | 1555 | |||
XM_005271660.1 | XP_005271717.1 | 9 | 1564 | |||
XM_005271661.1 | XP_005271718.1 | 8 | 1435 | |||
XM_005271664.1 | XP_005271721.1 | 7 | 1260 | |||
XM_006718901.2 | XP_006718964.1 | 10 | 1628 | |||
XM_011542972.1 | XP_011541274.1 | 8 | 1724 | |||
XM_011542973.1 | XP_011541275.1 | 9 | 1834 | |||
XM_011542975.1 | XP_011541277.1 | 9 | 1997 | |||
XM_011542976.1 | XP_011541278.1 | 8 | 1720 | |||
XM_017018268.1 | XP_016873757.1 | 10 | 1695 | |||
GenBank mRNA | AF027218.1 | - | 6 | 1101 | ||
AF027219.1 | - | 6 | 1105 | |||
AF155101.1 | - | 6 | 793 | |||
AK023751.1 | - | 7 | 1261 | |||
AK226047.1 | - | 8 | 1470 | |||
AL162031.1 | - | 6 | 1055 | |||
BC013382.2 | - | 9 | 1495 | 3508386 |