Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mndal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170853.1 | NP_001164324.1 | 459 | |||
GenBank mRNA | AK087931.1 | - | 2 | 288 | ||
AK136391.1 | - | 460 | ||||
FJ594740.1 | - | 2 | 358 | |||
FJ594741.1 | - | 367 |
Gene Symbol: Ifi203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF022371.1 | - | 520 | |||
AK169754.1 | - | 4 | 454 | |||
AK172303.1 | - | 513 | ||||
AK172505.1 | - | 534 | ||||
BC008167.1 | - | 562 | 3257492 |