Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnni2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017185.1 | NP_058881.1 | 7 | 526 | ||
GenBank mRNA | FQ216337.1 | - | 7 | 568 | ||
FQ217026.1 | - | 8 | 568 | |||
FQ217309.1 | - | 7 | 559 | |||
FQ217454.1 | - | 8 | 576 | |||
FQ217985.1 | - | 8 | 568 | |||
FQ218058.1 | - | 7 | 600 | |||
FQ223581.1 | - | 8 | 568 | |||
FQ223772.1 | - | 8 | 574 | |||
FQ224402.1 | - | 2 | 190 | |||
FQ224418.1 | - | 2 | 99 | |||
FQ224464.1 | - | 8 | 568 | |||
FQ224489.1 | - | 8 | 568 | |||
M73701.1 | - | 7 | 526 |