Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017266.2 | NP_058962.2 | 3 | 294 | ||
XM_006229233.2 | XP_006229295.1 | 3 | 365 | |||
XM_008759344.1 | XP_008757566.1 | 3 | 241 | |||
XM_008759345.1 | XP_008757567.1 | 3 | 241 | |||
GenBank mRNA | BC078862.1 | - | 3 | 294 | 7131609 | |
JQ173138.1 | - | 2 | 191 | |||
JQ173140.1 | - | 2 | 191 | |||
JQ173141.1 | - | 2 | 191 | |||
JQ409364.1 | - | 2 | 191 | |||
JQ409366.1 | - | 2 | 191 | |||
JQ409367.1 | - | 2 | 191 | |||
JQ409368.1 | - | 2 | 191 | |||
JQ409369.1 | - | 2 | 191 | |||
U07177.1 | - | 2 | 195 |