Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024142.1 | NP_001019313.1 | 3 | 844 | ||
GenBank mRNA | AK142389.1 | - | 3 | 839 | ||
AK150292.1 | - | 3 | 834 | |||
AK153358.1 | - | 3 | 834 | |||
BC052485.1 | - | 2 | 311 | |||
BC054731.1 | - | 3 | 834 | |||
BC062126.1 | - | 2 | 203 | |||
BC094565.1 | - | 2 | 628 | 6836944 |