Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017339.3 | NP_059035.3 | 2, 3 | 231 | ||
GenBank mRNA | AY557632.1 | - | 2, 3 | 226 | ||
S69329.1 | - | 2, 3 | 231 | |||
X53258.1 | - | 2, 3 | 334 |