Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136181.1 | NP_001129653.1 | 5 | 1069 | ||
GenBank mRNA | AK141935.1 | - | 5 | 1070 |