Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029368.1 | NP_083644.1 | 5 | 524 | ||
GenBank mRNA | AK006487.1 | - | 3 | 556 | ||
AK160565.1 | - | 5 | 525 | |||
BC099557.1 | - | 4 | 437 | 30916402 | ||
BC116881.1 | - | 3 | 721 | 40126200 | ||
BC119032.1 | - | 3 | 789 | 8733781 |