Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kidins220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081378.1 | NP_001074847.1 | 32 | 6966 | ||
GenBank mRNA | AK014150.1 | - | 1 | 202 | ||
AK122478.1 | - | 29 | 6559 | |||
AK151805.1 | - | 1 | 1649 | |||
BC030876.1 | - | 1 | 2142 | |||
BC044772.2 | - | 23 | 6085 | |||
BC046467.1 | - | 23 | 6085 | |||
BC059030.1 | - | 1 | 2394 | |||
BC094243.1 | - | 6 | 3462 | |||
KJ812113.1 | - | 9 | 3610 | |||
KJ812114.1 | - | 8 | 3577 | |||
KJ812115.1 | - | 7 | 3520 | |||
KJ812116.1 | - | 6 | 3487 | |||
KJ812117.1 | - | 6 | 3406 | |||
KJ812118.1 | - | 5 | 3373 |