Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019340.1 | NP_062213.1 | 4 | 431 | ||
XM_006238260.2 | XP_006238322.1 | 11 | 1014 | |||
XM_006238263.2 | XP_006238325.1 | 13 | 1189 | |||
XM_006238264.2 | XP_006238326.1 | 11 | 1016 | |||
XM_006238265.2 | XP_006238327.1 | 4 | 2631 | |||
GenBank mRNA | AB055153.1 | - | 4 | 431 | ||
BC085710.1 | - | 4 | 420 |