Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pclo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110797.1 | NP_001104267.1 | 407 | |||
NM_020098.1 | NP_064483.1 | 407 | ||||
GenBank mRNA | AF138789.2 | - | 407 | |||
AF227534.1 | - | 407 |