Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYL1 SIRNA Assays ›
Gene Symbol:
Myl1
Gene Name:
myosin, light chain 1
Gene Aliases:
MLC3-f, MLCf, Mcl3, Mlc1, Mlc3
Chromosome Location:
Chr.9: 73937820 - 73958480 on Build Rnor_6.0
UniGene ID:
Rn.84920
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
51173
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Myl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077656.1
NP_001071124.1
4
496
NM_020104.2
NP_064489.1
4
314
XM_008767294.1
XP_008765516.1
3
444
XM_008767295.1
XP_008765517.1
3
565
GenBank mRNA
CR755322.1
-
4
319
CR756356.1
-
4
314
FQ214608.1
-
4
501
FQ214631.1
-
4
483
FQ214638.1
-
4
591
FQ214656.1
-
4
332
FQ214675.1
-
4
590
FQ214712.1
-
4
504
FQ214723.1
-
4
504
FQ214727.1
-
4
504
FQ214742.1
-
4
504
FQ214939.1
-
4
501
FQ215002.1
-
4
590
FQ215050.1
-
4
504
FQ215083.1
-
4
501
FQ215086.1
-
4
334
FQ215201.1
-
4
501
FQ215229.1
-
4
595
FQ215325.1
-
4
590
FQ215340.1
-
4
332
FQ215418.1
-
4
590
FQ215435.1
-
4
501
FQ215492.1
-
4
483
FQ215500.1
-
3
1080
FQ215535.1
-
3
565
FQ215557.1
-
4
590
FQ215563.1
-
4
501
FQ215578.1
-
4
590
FQ215748.1
-
3
563
FQ215766.1
-
4
590
FQ215774.1
-
4
501
FQ215820.1
-
4
559
FQ215848.1
-
4
501
FQ215875.1
-
4
501
FQ215911.1
-
4
595
FQ215933.1
-
4
504
FQ216034.1
-
4
504
FQ216060.1
-
4
590
FQ216109.1
-
4
501
FQ216124.1
-
4
595
FQ216181.1
-
4
483
FQ216192.1
-
4
590
FQ216213.1
-
5
498
FQ216218.1
-
4
501
FQ216229.1
-
4
771
FQ216238.1
-
4
590
FQ216308.1
-
4
332
FQ216407.1
-
4
672
FQ216452.1
-
4
501
FQ216455.1
-
4
505
FQ216484.1
-
4
483
FQ216496.1
-
4
504
FQ216525.1
-
4
504
FQ216543.1
-
4
501
FQ216556.1
-
4
590
FQ216567.1
-
4
501
FQ216631.1
-
4
504
FQ216654.1
-
4
588
FQ216704.1
-
4
504
FQ216737.1
-
4
587
FQ216738.1
-
4
332
FQ217074.1
-
4
331
FQ217103.1
-
4
332
FQ217157.1
-
3
307
FQ217192.1
-
4
332
FQ217218.1
-
4
332
FQ217243.1
-
4
331
FQ217303.1
-
4
483
FQ217323.1
-
4
332
FQ217387.1
-
4
332
FQ217394.1
-
4
434
FQ217427.1
-
4
337
FQ217481.1
-
4
332
FQ217524.1
-
4
332
FQ217753.1
-
4
337
FQ217756.1
-
4
332
FQ217889.1
-
4
332
FQ217973.1
-
4
337
FQ217979.1
-
4
332
FQ218029.1
-
4
332
FQ218049.1
-
4
332
FQ223584.1
-
4
590
FQ223632.1
-
4
504
FQ223641.1
-
4
590
FQ223687.1
-
4
332
FQ223755.1
-
4
500
FQ223779.1
-
4
332
FQ223827.1
-
4
504
FQ223880.1
-
4
402
FQ223949.1
-
4
330
FQ223984.1
-
4
332
FQ224047.1
-
4
332
FQ224050.1
-
4
332
FQ224087.1
-
4
332
FQ224155.1
-
4
337
FQ224171.1
-
4
501
FQ224172.1
-
4
332
FQ224212.1
-
4
484
FQ224254.1
-
4
332
FQ224277.1
-
4
504
FQ224321.1
-
4
336
FQ224386.1
-
4
337
FQ224411.1
-
4
332
FQ224420.1
-
4
332
FQ224452.1
-
4
407
FQ224519.1
-
4
501
FQ224609.1
-
5
360
FQ224663.1
-
4
332
FQ224726.1
-
4
434
FQ224796.1
-
4
332
FQ224799.1
-
4
337
FQ224867.1
-
4
590
FQ224927.1
-
4
590
FQ224931.1
-
4
501
FQ224934.1
-
4
499
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cardiac muscle contraction
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0