Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022958.2 | NP_075247.1 | 2 | 114 | ||
XM_006254488.2 | XP_006254550.1 | 2 | 195 | |||
GenBank mRNA | AJ006710.1 | - | 2 | 96 | ||
BC061981.1 | - | 2 | 114 | 5597998 |