Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024353.1 | NP_077329.1 | 4, 5 | 359 | ||
XM_006235083.2 | XP_006235145.1 | 4, 5 | 396 | |||
XM_006235085.2 | XP_006235147.1 | 4, 5 | 257 | |||
XM_006235088.2 | XP_006235150.1 | 3, 4 | 2510 | |||
XM_006235089.2 | XP_006235151.1 | 4, 5 | 3083 | |||
XM_006235094.2 | XP_006235156.1 | 4, 5 | 396 | |||
XM_008762248.1 | XP_008760470.1 | 4, 5 | 2629 | |||
XM_008762249.1 | XP_008760471.1 | 4, 5 | 2629 | |||
XM_008762250.1 | XP_008760472.1 | 4, 5 | 2629 | |||
XM_008762251.1 | XP_008760473.1 | 4, 5 | 2629 | |||
GenBank mRNA | AF031370.1 | - | 2, 3 | 129 | ||
L15556.1 | - | 4, 5 | 359 | |||
L18962.1 | - | 4, 5 | 262 | |||
U57836.1 | - | 4, 5 | 262 |