Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031143.1 | NP_112405.1 | 1, 2 | 349 | ||
GenBank mRNA | D78588.1 | - | 1, 2 | 349 |