Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031721.1 | NP_113909.1 | 1, 2 | 520 | ||
GenBank mRNA | AF179370.1 | - | 1, 2 | 520 | ||
BC081767.1 | - | 1, 2 | 545 | 7121959 |