Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053307.2 | NP_445759.1 | 2 | 201 | ||
XM_008770745.1 | XP_008768967.1 | 2 | 572 | |||
GenBank mRNA | AY005464.1 | - | 2 | 191 | ||
BC087009.1 | - | 2 | 201 | 7107102 | ||
DQ989019.1 | - | 3 | 233 | |||
DQ989020.1 | - | 3 | 236 |