Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herpud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053523.1 | NP_445975.1 | 2 | 263 | ||
XM_006255133.2 | XP_006255195.1 | 2 | 579 | |||
GenBank mRNA | AB033771.1 | - | 2 | 263 | ||
BC061849.1 | - | 2 | 285 | 5621877 |