Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053811.3 | NP_446263.2 | 6, 7 | 929 | ||
XM_006245893.2 | XP_006245955.1 | 5, 6 | 1302 | |||
GenBank mRNA | AF259898.1 | - | 6, 7 | 788 | ||
AF294257.1 | - | 6, 7 | 1016 |