Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053820.2 | NP_446272.2 | 2, 3 | 581 | ||
XM_006246117.2 | XP_006246179.1 | 2, 3 | 2162 | |||
XM_006246118.2 | XP_006246180.1 | 2, 3 | 2162 | |||
XM_006246120.2 | XP_006246182.1 | 3, 4 | 1916 | |||
XM_008767631.1 | XP_008765853.1 | 2, 3 | 516 | |||
XM_008767632.1 | XP_008765854.1 | 2, 3 | 930 | |||
GenBank mRNA | FQ175272.1 | - | 2, 3 | 581 | ||
L24051.1 | - | 2, 3 | 323 |