Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CARD8 SIRNA Assays ›
          Gene Symbol:
          CARD8
          Gene Name:
          caspase recruitment domain family member 8
          Gene Aliases:
          CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
          Chromosome Location:
          Chr.19: 48203148 - 48256269 on Build GRCh38
          UniGene ID:
          Hs.446146
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 5620

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CARD8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001184900.1 NP_001171829.1 11 4085
          NM_001184901.1 NP_001171830.1 11 4032
          NM_001184902.1 NP_001171831.1 13 4306
          NM_001184903.1 NP_001171832.1 11 4040
          NM_014959.3 NP_055774.2 12 4143
          NR_033678.1 10 4002
          NR_033679.1 10 4044
          NR_033680.1 9 3860
          XM_006723091.3 XP_006723154.1 13 4296
          XM_006723092.3 XP_006723155.1 15 4834
          XM_006723093.3 XP_006723156.1 13 4617
          XM_006723094.3 XP_006723157.1 13 4276
          XM_006723095.3 XP_006723158.1 13 4263
          XM_006723096.3 XP_006723159.1 14 4685
          XM_006723097.3 XP_006723160.1 12 4468
          XM_006723098.3 XP_006723161.1 12 4255
          XM_006723099.3 XP_006723162.1 11 4141
          XM_006723102.3 XP_006723165.1 11 4098
          XM_006723104.3 XP_006723167.1 12 4155
          XM_006723106.3 XP_006723169.1 12 4486
          XM_006723107.3 XP_006723170.1 11 4011
          XM_006723109.3 XP_006723172.1 11 4013
          XM_011526641.2 XP_011524943.1 16 5141
          XM_011526642.2 XP_011524944.1 15 5036
          XM_011526643.2 XP_011524945.1 14 4723
          XM_011526644.2 XP_011524946.1 12 4308
          XM_011526646.2 XP_011524948.1 13 4782
          XM_011526647.2 XP_011524949.1 12 4671
          XM_011526648.2 XP_011524950.1 13 4788
          XM_011526653.2 XP_011524955.1 12 4676
          XM_017026482.1 XP_016881971.1 13 4621
          XM_017026483.1 XP_016881972.1 13 4574
          XM_017026484.1 XP_016881973.1 13 4582
          XM_017026486.1 XP_016881975.1 11 4138
          XM_017026487.1 XP_016881976.1 10 4507
          XM_017026488.1 XP_016881977.1 14 4723
          XM_017026492.1 XP_016881981.1 12 4448
          XM_017026493.1 XP_016881982.1 11 4340
          XM_017026494.1 XP_016881983.1 12 4954
          XM_017026495.1 XP_016881984.1 11 4096
          XM_017026496.1 XP_016881985.1 11 4016
          XM_017026497.1 XP_016881986.1 10 3956
          XM_017026498.1 XP_016881987.1 10 3953
          XM_017026499.1 XP_016881988.1 9 5726
          GenBank mRNA AB023172.1 - 10 4037
          AF322184.1 - 10 4037

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of interleukin-1 beta secretion
          protein binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          NACHT domain binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0