Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080786.1 | NP_542964.1 | 11, 12 | 1820 | ||
XM_006229721.2 | XP_006229783.1 | 11, 12 | 2129 | |||
XM_006229722.2 | XP_006229784.1 | 11, 12 | 2126 | |||
XM_006229723.2 | XP_006229785.1 | 11, 12 | 3129 | |||
XM_006229724.2 | XP_006229786.1 | 11, 12 | 3100 | |||
XM_006229725.2 | XP_006229787.1 | 11, 12 | 3085 | |||
XM_006229727.2 | XP_006229789.1 | 11, 12 | 3097 | |||
XM_006229728.2 | XP_006229790.1 | 11, 12 | 3082 | |||
XM_008759668.1 | XP_008757890.1 | 12, 13 | 1919 | |||
GenBank mRNA | AF169409.1 | - | 11, 12 | 1820 | ||
AF169410.1 | - | 11, 12 | 1780 |