Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133593.2 | NP_598277.2 | 2, 3 | 311 | ||
XM_006251629.1 | XP_006251691.1 | 2, 3 | 838 | |||
XM_006251630.2 | XP_006251692.1 | 2, 3 | 357 | |||
GenBank mRNA | BC070925.1 | - | 2, 3 | 311 | 7096586 | |
L07073.1 | - | 2, 3 | 324 |