Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147209.2 | NP_671742.1 | 3 | 381 | ||
GenBank mRNA | AF525687.1 | - | 3 | 257 | ||
BC062083.1 | - | 3 | 381 | 5599833 |