Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172038.1 | NP_742035.1 | 7 | 566 | ||
GenBank mRNA | U86635.1 | - | 7 | 566 |
Gene Symbol: LOC100912430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749378.3 | XP_003749426.1 | 7 | 702 |