Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ambra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134341.1 | NP_001127813.1 | 7 | 2133 | ||
XM_008762029.1 | XP_008760251.1 | 7 | 2320 | |||
XM_008762030.1 | XP_008760252.1 | 7 | 2037 | |||
XM_008762031.1 | XP_008760253.1 | 6 | 1894 | |||
XM_008762032.1 | XP_008760254.1 | 6 | 1894 | |||
XM_008762033.1 | XP_008760255.1 | 7 | 1621 | |||
XM_008762034.1 | XP_008760256.1 | 7 | 1621 | |||
GenBank mRNA | AB027149.1 | - | 1 | 209 |