Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACTA1 SIRNA Assays ›
          Gene Symbol:
          Acta1
          Gene Name:
          actin, alpha 1, skeletal muscle
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 56674072 - 56677084 on Build Rnor_6.0
          UniGene ID:
          Rn.82732
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 59691

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Acta1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019212.2 NP_062085.1 3 417
          GenBank mRNA BC061974.1 - 3 417 5623909
          FQ214583.1 - 3 426
          FQ214585.1 - 3 428
          FQ214598.1 - 3 426
          FQ214600.1 - 3 426
          FQ214603.1 - 3 426
          FQ214623.1 - 3 426
          FQ214640.1 - 3 426
          FQ214659.1 - 3 428
          FQ214676.1 - 3 426
          FQ214680.1 - 3 426
          FQ214697.1 - 3 426
          FQ214743.1 - 3 426
          FQ214756.1 - 3 426
          FQ214761.1 - 3 425
          FQ214779.1 - 3 426
          FQ214785.1 - 3 424
          FQ214786.1 - 3 426
          FQ214807.1 - 3 426
          FQ214814.1 - 3 426
          FQ214822.1 - 3 426
          FQ214831.1 - 3 426
          FQ214832.1 - 3 426
          FQ214839.1 - 3 429
          FQ214841.1 - 3 426
          FQ214842.1 - 3 426
          FQ214857.1 - 3 430
          FQ214858.1 - 3 426
          FQ214859.1 - 3 426
          FQ214860.1 - 3 426
          FQ214862.1 - 3 426
          FQ214866.1 - 3 426
          FQ214883.1 - 3 425
          FQ214904.1 - 3 426
          FQ214905.1 - 3 425
          FQ214916.1 - 3 426
          FQ214921.1 - 3 426
          FQ214936.1 - 3 426
          FQ214948.1 - 3 426
          FQ214949.1 - 3 426
          FQ214957.1 - 3 426
          FQ214969.1 - 3 426
          FQ214975.1 - 3 428
          FQ214978.1 - 3 426
          FQ214987.1 - 3 426
          FQ214990.1 - 3 426
          FQ214993.1 - 3 426
          FQ215012.1 - 3 426
          FQ215025.1 - 3 426
          FQ215038.1 - 3 426
          FQ215044.1 - 3 426
          FQ215058.1 - 3 426
          FQ215089.1 - 3 426
          FQ215112.1 - 3 426
          FQ215128.1 - 3 428
          FQ215136.1 - 3 426
          FQ215149.1 - 3 426
          FQ215153.1 - 3 426
          FQ215158.1 - 3 424
          FQ215173.1 - 3 426
          FQ215185.1 - 3 425
          FQ215204.1 - 3 424
          FQ215212.1 - 3 426
          FQ215215.1 - 3 426
          FQ215225.1 - 3 426
          FQ215241.1 - 3 426
          FQ215249.1 - 3 427
          FQ215253.1 - 3 426
          FQ215256.1 - 3 426
          FQ215275.1 - 3 426
          FQ215277.1 - 3 427
          FQ215280.1 - 3 426
          FQ215282.1 - 3 426
          FQ215298.1 - 3 428
          FQ215324.1 - 3 426
          FQ215333.1 - 3 426
          FQ215342.1 - 3 426
          FQ215346.1 - 3 426
          FQ215366.1 - 3 426
          FQ215380.1 - 3 426
          FQ215381.1 - 3 426
          FQ215393.1 - 3 426
          FQ215396.1 - 3 426
          FQ215398.1 - 3 426
          FQ215402.1 - 3 426
          FQ215403.1 - 3 426
          FQ215405.1 - 3 426
          FQ215414.1 - 3 428
          FQ215424.1 - 3 426
          FQ215431.1 - 3 426
          FQ215433.1 - 3 425
          FQ215434.1 - 3 428
          FQ215438.1 - 3 426
          FQ215446.1 - 3 426
          FQ215449.1 - 3 428
          FQ215454.1 - 3 426
          FQ215464.1 - 3 426
          FQ215465.1 - 3 426
          FQ215468.1 - 3 426
          FQ215470.1 - 3 426
          FQ215474.1 - 3 426
          FQ215483.1 - 3 426
          FQ215488.1 - 3 426
          FQ215518.1 - 3 435
          FQ215523.1 - 3 426
          FQ215537.1 - 3 428
          FQ215554.1 - 3 426
          FQ215567.1 - 3 426
          FQ215572.1 - 3 426
          FQ215593.1 - 3 426
          FQ215618.1 - 3 426
          FQ215628.1 - 3 426
          FQ215631.1 - 1 40
          FQ215655.1 - 3 424
          FQ215673.1 - 3 426
          FQ215674.1 - 3 426
          FQ215690.1 - 3 426
          FQ215692.1 - 3 426
          FQ215785.1 - 3 426
          FQ215790.1 - 3 428
          FQ215799.1 - 3 426
          FQ215809.1 - 3 426
          FQ215813.1 - 3 426
          FQ215835.1 - 3 426
          FQ215836.1 - 3 426
          FQ215839.1 - 3 426
          FQ215840.1 - 3 426
          FQ215841.1 - 3 426
          FQ215851.1 - 3 426
          FQ215852.1 - 3 424
          FQ215864.1 - 3 427
          FQ215866.1 - 3 428
          FQ215888.1 - 3 425
          FQ215893.1 - 3 427
          FQ215896.1 - 3 426
          FQ215907.1 - 3 428
          FQ215924.1 - 1 118
          FQ215950.1 - 3 426
          FQ215964.1 - 3 426
          FQ215972.1 - 3 426
          FQ216016.1 - 3 426
          FQ216035.1 - 3 426
          FQ216043.1 - 3 426
          FQ216050.1 - 3 426
          FQ216066.1 - 3 424
          FQ216139.1 - 3 426
          FQ216168.1 - 3 424
          FQ216196.1 - 3 426
          FQ216224.1 - 3 426
          FQ216246.1 - 3 426
          FQ216276.1 - 3 426
          FQ216278.1 - 3 428
          FQ216287.1 - 3 424
          FQ216291.1 - 3 426
          FQ216293.1 - 3 426
          FQ216305.1 - 3 426
          FQ216329.1 - 3 426
          FQ216333.1 - 3 426
          FQ216350.1 - 3 426
          FQ216352.1 - 3 426
          FQ216360.1 - 3 426
          FQ216403.1 - 3 426
          FQ216430.1 - 3 426
          FQ216509.1 - 3 424
          FQ216586.1 - 3 426
          FQ216603.1 - 3 426
          FQ216606.1 - 3 426
          FQ216607.1 - 3 422
          FQ216610.1 - 3 424
          FQ216620.1 - 3 426
          FQ216635.1 - 3 424
          FQ216639.1 - 3 426
          FQ216644.1 - 3 426
          FQ216664.1 - 3 424
          FQ216675.1 - 3 426
          FQ216676.1 - 3 426
          FQ216692.1 - 3 426
          FQ216703.1 - 3 426
          FQ216713.1 - 3 426
          FQ216721.1 - 3 426
          FQ216723.1 - 3 426
          FQ216725.1 - 3 422
          FQ216762.1 - 3 424
          FQ216767.1 - 3 426
          FQ216776.1 - 3 426
          FQ216777.1 - 3 426
          FQ216779.1 - 3 428
          FQ216799.1 - 3 424
          FQ216801.1 - 3 426
          FQ216814.1 - 3 420
          FQ216819.1 - 3 424
          FQ216916.1 - 3 426
          FQ216918.1 - 1 69
          FQ216961.1 - 3 426
          FQ216994.1 - 3 426
          FQ217061.1 - 3 428
          FQ217073.1 - 3 429
          FQ217091.1 - 3 425
          FQ217114.1 - 3 426
          FQ217122.1 - 3 426
          FQ217167.1 - 3 426
          FQ217173.1 - 3 426
          FQ217188.1 - 3 426
          FQ217194.1 - 3 426
          FQ217273.1 - 3 425
          FQ217280.1 - 3 546
          FQ217327.1 - 3 426
          FQ217329.1 - 3 426
          FQ217347.1 - 3 426
          FQ217353.1 - 3 426
          FQ217367.1 - 3 426
          FQ217397.1 - 3 426
          FQ217441.1 - 3 426
          FQ217443.1 - 3 428
          FQ217475.1 - 3 426
          FQ217525.1 - 3 426
          FQ217535.1 - 3 426
          FQ217572.1 - 3 426
          FQ217593.1 - 3 426
          FQ217602.1 - 3 426
          FQ217615.1 - 3 426
          FQ217629.1 - 3 426
          FQ217641.1 - 3 423
          FQ217762.1 - 3 426
          FQ217826.1 - 3 426
          FQ217893.1 - 3 426
          FQ217905.1 - 3 426
          FQ217908.1 - 3 426
          FQ217919.1 - 3 426
          FQ217937.1 - 3 426
          FQ217969.1 - 3 428
          FQ217990.1 - 3 426
          FQ218019.1 - 3 426
          FQ218034.1 - 3 426
          FQ223563.1 - 3 426
          FQ223681.1 - 3 428
          FQ223686.1 - 3 426
          FQ223702.1 - 3 426
          FQ223727.1 - 1 124
          FQ223898.1 - 3 426
          FQ223926.1 - 3 426
          FQ223929.1 - 3 426
          FQ224009.1 - 3 426
          FQ224082.1 - 3 426
          FQ224141.1 - 3 427
          FQ224167.1 - 3 426
          FQ224359.1 - 3 426
          FQ224377.1 - 3 426
          FQ224419.1 - 3 426
          FQ224552.1 - 3 426
          FQ224691.1 - 3 424
          FQ224777.1 - 3 426
          FQ224840.1 - 3 424
          FQ224842.1 - 3 426
          FQ224869.1 - 3 426
          FQ224886.1 - 3 426
          FQ224890.1 - 3 424
          FQ224902.1 - 3 426
          FQ224929.1 - 3 424
          FQ224936.1 - 3 426
          FQ224942.1 - 3 428
          FQ224948.1 - 3 428
          FQ224962.1 - 3 426
          FQ224971.1 - 3 426

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001272041, NM_009606, M12866, BC014877, X03766, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          response to mechanical stimulus
          response to extracellular stimulus
          response to lithium ion
          cell growth
          skeletal muscle thin filament assembly
          skeletal muscle fiber adaptation
          response to steroid hormone
          skeletal muscle fiber development
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b8d7ffddb-2cdg5:80/100.66.130.62:80.
          git-commit: 44eaffc35a31f29f64b20d0c2dd51bfa522c97f5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0