Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022503.2 | NP_071948.1 | 2 | 172 | ||
GenBank mRNA | BC166880.1 | - | 2 | 140 | 6888806 | |
BG668415.1 | - | 2 | 179 | |||
FQ209801.1 | - | 2 | 167 | |||
FQ209847.1 | - | 2 | 139 | |||
FQ210551.1 | - | 2 | 139 | |||
FQ211276.1 | - | 2 | 140 | |||
FQ214317.1 | - | 2 | 148 | |||
FQ217361.1 | - | 2 | 139 | |||
FQ221839.1 | - | 2 | 144 | |||
FQ223968.1 | - | 2 | 163 | |||
FQ228537.1 | - | 2 | 211 | |||
FQ229029.1 | - | 2 | 141 | |||
FQ229035.1 | - | 2 | 173 | |||
FQ230547.1 | - | 2 | 200 | |||
X54080.1 | - | 2 | 146 |
Gene Symbol: Cox7a2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751367.3 | XP_003751415.1 | 1 | 123 |