Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252572.1 | NP_001239501.1 | 9 | 958 | ||
NM_007803.5 | NP_031829.2 | 9 | 958 | |||
GenBank mRNA | AK084249.1 | - | 9 | 768 | ||
AK146856.1 | - | 9 | 765 | |||
AK148010.1 | - | 9 | 761 | |||
AK165555.1 | - | 9 | 761 | |||
AK168699.1 | - | 9 | 759 | |||
AK168871.1 | - | 9 | 759 | |||
AK168945.1 | - | 9 | 769 | |||
AK177866.1 | - | 4 | 263 | |||
AK184325.1 | - | 4 | 263 | |||
AK189485.1 | - | 4 | 263 | |||
AK206447.1 | - | 4 | 263 | |||
AK216063.1 | - | 4 | 263 | |||
AK216633.1 | - | 4 | 263 | |||
AK219085.1 | - | 4 | 263 | |||
BC011434.1 | - | 9 | 751 | 3981559 | ||
BC117820.1 | - | 8 | 660 | |||
U03184.1 | - | 9 | 747 |