Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008960.2 | NP_032986.1 | 2 | 1016 | ||
GenBank mRNA | AK076980.1 | - | 2 | 974 | ||
AK088717.1 | - | 2 | 995 | |||
AK164302.1 | - | 2 | 999 | |||
AK208703.1 | - | 2 | 248 | |||
BC021445.1 | - | 2 | 365 | 5038272 | ||
U92437.1 | - | 2 | 1096 |