Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205313.1 | NP_001192242.1 | 5 | 637 | ||
NM_001205314.1 | NP_001192243.1 | 5 | 673 | |||
NM_009283.4 | NP_033309.3 | 5 | 688 | |||
GenBank mRNA | AK018544.1 | - | 5 | 639 | ||
AK039458.1 | - | 5 | 689 | |||
AK046517.1 | - | 5 | 637 | |||
AK082706.1 | - | 5 | 670 | |||
AK084855.1 | - | 5 | 622 | |||
AK088874.1 | - | 5 | 557 | |||
AK157263.1 | - | 5 | 537 | |||
AK159887.1 | - | 5 | 672 | |||
BC004808.1 | - | 5 | 624 | 3587831 | ||
U06924.1 | - | 3 | 342 |