Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010353.2 | NP_034483.1 | 1 | 743 | ||
GenBank mRNA | AK138750.1 | - | 1 | 744 | ||
BC113148.1 | - | 1 | 737 | 40054435 | ||
D87326.1 | - | 1 | 743 |