Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110498.1 | NP_001103968.1 | 6 | 827 | ||
NM_010509.2 | NP_034639.2 | 6 | 827 | |||
GenBank mRNA | AF013274.1 | - | 6 | 605 | ||
AF013486.1 | - | 6 | 699 | |||
AK003185.1 | - | 6 | 761 | |||
AK010952.1 | - | 6 | 778 | |||
AK139169.1 | - | 6 | 807 | |||
AK150386.1 | - | 6 | 761 | |||
AK150597.1 | - | 6 | 740 | |||
AK151474.1 | - | 6 | 755 | |||
AK151493.1 | - | 6 | 740 | |||
AK151777.1 | - | 6 | 755 | |||
AK152878.1 | - | 6 | 761 | |||
AK153317.1 | - | 6 | 761 | |||
BC003743.1 | - | 5 | 522 | |||
BC071225.1 | - | 5 | 525 | 4187603 | ||
Y09813.1 | - | 6 | 582 | |||
Y09864.1 | - | 6 | 717 | |||
Y09865.1 | - | 6 | 717 |