Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: I830012O16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005858.3 | NP_001005858.2 | 127 | |||
GenBank mRNA | AK077243.1 | - | 116 | |||
AK150632.1 | - | 2 | 374 | |||
AK151442.1 | - | 2 | 374 | |||
AK152620.1 | - | 2 | 374 | |||
CA576017.1 | - | 127 |
Gene Symbol: Ifit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010501.2 | NP_034631.1 | 2 | 161 | ||
GenBank mRNA | AK077459.1 | - | 2 | 159 | ||
AK151032.1 | - | 2 | 161 | |||
AK153226.1 | - | 2 | 162 | |||
BC003804.1 | - | 2 | 127 | 3487345 | ||
BC089563.1 | - | 2 | 159 | 30317428 | ||
CT010266.1 | - | 1 | 90 | |||
L32974.1 | - | 2 | 142 |