Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010509.2 | NP_034639.2 | 9 | 2111 | ||
GenBank mRNA | AF013274.1 | - | 9 | 1889 | ||
AK139169.1 | - | 9 | 2091 | |||
AK150386.1 | - | 9 | 2045 | |||
AK150597.1 | - | 9 | 2024 | |||
AK151474.1 | - | 9 | 2039 | |||
AK151493.1 | - | 9 | 2024 | |||
AK151777.1 | - | 9 | 2040 | |||
AK152878.1 | - | 9 | 2045 | |||
AK153317.1 | - | 9 | 2045 | |||
BC071225.1 | - | 8 | 1809 | 4187603 | ||
Y09864.1 | - | 8 | 1872 | |||
Y09865.1 | - | 9 | 2001 |