Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161769.1 | NP_001155241.1 | 3 | 473 | ||
NM_001161770.1 | NP_001155242.1 | 2 | 605 | |||
NM_010723.3 | NP_034853.1 | 3 | 760 | |||
GenBank mRNA | AF074600.1 | - | 2 | 321 | ||
AF096996.1 | - | 3 | 489 | |||
AF102817.1 | - | 3 | 390 | |||
AK080534.1 | - | 3 | 408 | |||
AK138278.1 | - | 2 | 606 | |||
AK159590.1 | - | 3 | 687 | |||
BC003488.1 | - | 2 | 472 | 2811636 | ||
BC004661.1 | - | 3 | 413 | |||
BC010278.1 | - | 3 | 503 | 3589532 | ||
BC100382.1 | - | 3 | 365 | 6487478 |