Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286181.1 | NP_001273110.1 | 8 | 2259 | ||
NM_001286182.1 | NP_001273111.1 | 8 | 2216 | |||
NM_010728.3 | NP_034858.2 | 7 | 2636 | |||
GenBank mRNA | AK135213.1 | - | 7 | 1767 | ||
AK141995.1 | - | 7 | 2522 | |||
AK159199.1 | - | 7 | 2439 | |||
AK159241.1 | - | 7 | 2415 | |||
AK159263.1 | - | 7 | 2476 | |||
AK159368.1 | - | 7 | 2398 | |||
AK164634.1 | - | 8 | 2217 | |||
AK166279.1 | - | 7 | 2343 | |||
AK166386.1 | - | 2479 | ||||
AK166442.1 | - | 2635 | ||||
BC018439.1 | - | 7 | 2487 | 2655752 | ||
M65142.1 | - | 7 | 2404 | |||
M65143.1 | - | 4 | 1262 |