Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159717.1 | NP_001153189.1 | 5 | 348 | ||
NM_001159718.1 | NP_001153190.1 | 4 | 321 | |||
NM_001159719.1 | NP_001153191.1 | 5 | 396 | |||
NM_010891.2 | NP_035021.1 | 5 | 352 | |||
GenBank mRNA | AK028072.1 | - | 5 | 334 | ||
AK146616.1 | - | 5 | 349 | |||
AK151591.1 | - | 5 | 336 | |||
AK169852.1 | - | 4 | 371 | |||
AK171331.1 | - | 5 | 331 | |||
BB849650.1 | - | 5 | 322 | |||
BC138636.1 | - | 4 | 289 | 8861656 | ||
BC138637.1 | - | 4 | 289 | 8861657 | ||
BY330456.1 | - | 4 | 297 | |||
D49382.2 | - | 5 | 307 |