Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013663.5 | NP_038691.1 | 4 | 490 | ||
GenBank mRNA | AK009895.1 | - | 4 | 478 | ||
AK011657.1 | - | 4 | 478 | |||
AK078891.1 | - | 4 | 478 | |||
AK088122.1 | - | 4 | 476 | |||
AK134177.1 | - | 4 | 478 | |||
AK135298.1 | - | 4 | 478 | |||
AK141440.1 | - | 3 | 2628 | |||
AK145089.1 | - | 4 | 479 | |||
AK145600.1 | - | 4 | 478 | |||
AK145715.1 | - | 4 | 478 | |||
AK146010.1 | - | 4 | 478 | |||
AK146253.1 | - | 4 | 476 | |||
AK146610.1 | - | 4 | 478 | |||
AK150912.1 | - | 4 | 478 | |||
AK152411.1 | - | 5 | 934 | |||
AK152704.1 | - | 4 | 479 | |||
AK152779.1 | - | 5 | 934 | |||
AK153073.1 | - | 5 | 934 | |||
AK153209.1 | - | 4 | 478 | |||
AK154046.1 | - | 4 | 478 | |||
BC068111.1 | - | 4 | 470 | 5710934 | ||
BC071196.1 | - | 4 | 452 | 30441615 | ||
BC083316.1 | - | 4 | 462 | 6818630 | ||
X53824.1 | - | 4 | 423 |