Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013765.2 | NP_038793.2 | 2 | 188 | ||
GenBank mRNA | AK008301.1 | - | 2 | 184 | ||
AK010689.2 | - | 2 | 183 | |||
AK012722.1 | - | 2 | 186 | |||
AK018702.1 | - | 2 | 184 | |||
AK152132.1 | - | 2 | 184 | |||
AV585706.1 | - | 1 | 142 | |||
BC036987.1 | - | 1 | 174 | 4987659 | ||
BC081452.1 | - | 1 | 161 | 6411109 | ||
BC100456.1 | - | 2 | 175 | 6541446 | ||
BU530648.1 | - | 2 | 189 | |||
U67770.1 | - | 1 | 165 |