Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277932.1 | NP_001264861.1 | 2 | 545 | ||
NM_015733.5 | NP_056548.2 | 2 | 545 | |||
GenBank mRNA | AB019600.1 | - | 2 | 302 | ||
AB019601.1 | - | 2 | 302 | |||
AK085095.1 | - | 2 | 334 | |||
AK085152.1 | - | 2 | 301 | |||
AK085561.1 | - | 2 | 320 | |||
AK154495.1 | - | 2 | 315 | |||
AK156500.1 | - | 2 | 448 | |||
AK162863.1 | - | 2 | 325 | |||
BC056372.1 | - | 2 | 327 | 6834067 | ||
BC056447.1 | - | 2 | 545 | 5721188 | ||
CT010400.1 | - | 2 | 302 |