Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289521.1 | NP_001276450.1 | 1, 2 | 262 | ||
NM_015753.4 | NP_056568.2 | 2, 3 | 599 | |||
GenBank mRNA | AF033116.1 | - | 1, 2 | 114 | ||
AK012377.1 | - | 1, 2 | 70 | |||
AK031541.1 | - | 3, 4 | 696 | |||
AK122312.1 | - | 2, 3 | 172 | |||
AK135557.1 | - | 2, 3 | 409 | |||
AK141157.1 | - | 1, 2 | 230 | |||
AK157561.1 | - | 2, 3 | 322 | |||
BC060699.1 | - | 2, 3 | 336 | 6408845 | ||
CB245498.1 | - | 2, 3 | 599 | |||
CJ130482.1 | - | 1, 2 | 262 |