Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PARK2 SIRNA Assays ›
          Gene Symbol:
          Park2
          Gene Name:
          Parkinson disease (autosomal recessive, juvenile) 2, parkin
          Gene Aliases:
          PRKN
          Chromosome Location:
          Chr.17: 10840384 - 12063370 on Build GRCm38
          UniGene ID:
          Mm.311110
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 72368

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Park2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016694.3 NP_057903.1 2 188
          GenBank mRNA AB019558.1 - 2 159
          AF250293.1 - 2 188
          AF250294.1 - 2 188
          AF250295.1 - 2 188
          BC113204.1 - 2 171 40047891
          FJ555555.1 - 1 159
          FJ555556.1 - 2 159
          FJ555557.1 - 2 159
          GU272056.1 - 1 159

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_020093, GU322363, AB039878, AF168004, AF381280, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          mitophagy
          negative regulation of protein phosphorylation
          synaptic transmission, dopaminergic
          startle response
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein monoubiquitination
          autophagy
          response to unfolded protein
          learning
          locomotory behavior
          adult locomotory behavior
          proteasomal protein catabolic process
          regulation of autophagy
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of mitochondrial fusion
          regulation of mitochondrion organization
          protein ubiquitination
          protein metabolic process
          regulation of protein ubiquitination
          negative regulation of actin filament bundle assembly
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of glucokinase activity
          response to endoplasmic reticulum stress
          synaptic transmission, glutamatergic
          regulation of dopamine metabolic process
          norepinephrine metabolic process
          dopamine metabolic process
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of DNA binding
          negative regulation of neuron apoptotic process
          cellular protein catabolic process
          negative regulation by host of viral genome replication
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of insulin secretion
          regulation of neurotransmitter secretion
          modulation of synaptic transmission
          protein stabilization
          positive regulation of neurotransmitter uptake
          dopamine uptake involved in synaptic transmission
          mitochondrion localization
          protein autoubiquitination
          regulation of mitochondrial membrane potential
          negative regulation of cell death
          protein K63-linked ubiquitination
          protein localization to mitochondrion
          aggresome assembly
          protein K48-linked ubiquitination
          protein K11-linked ubiquitination
          protein K6-linked ubiquitination
          negative regulation of release of cytochrome c from mitochondria
          cellular response to toxic substance
          mitophagy in response to mitochondrial depolarization
          regulation of cellular response to oxidative stress
          negative regulation of neuron death
          positive regulation of proteasomal protein catabolic process
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          negative regulation of primary amine oxidase activity
          positive regulation of protein linear polyubiquitination
          negative regulation of oxidative stress-induced neuron death
          regulation of protein targeting to mitochondrion
          positive regulation of tumor necrosis factor-mediated signaling pathway
          negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          negative regulation of endoplasmic reticulum stress-induced neuron intrinsic apoptotic signaling pathway
          positive regulation of dendrite extension
          negative regulation of spontaneous neurotransmitter secretion
          regulation of reactive oxygen species metabolic process
          negative regulation of reactive oxygen species metabolic process
          G-protein coupled receptor binding
          actin binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          tubulin binding
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          enzyme binding
          kinase binding
          protein kinase binding
          PDZ domain binding
          Hsp70 protein binding
          heat shock protein binding
          ubiquitin conjugating enzyme binding
          ubiquitin protein ligase binding
          protein complex binding
          identical protein binding
          histone deacetylase binding
          ubiquitin binding
          phospholipase binding
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          chaperone binding
          ubiquitin protein ligase activity
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity
          cullin family protein binding
          ubiquitin-specific protease binding
          F-box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0