Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289653.1 | NP_001276582.1 | 12, 13 | 1240 | ||
GenBank mRNA | AK035019.1 | - | 10, 11 | 1093 | ||
AK039790.1 | - | 1304 | ||||
AK048913.1 | - | 1511 | ||||
BC051968.1 | - | 11, 12 | 1208 | 6401465 | ||
U59230.1 | - | 12, 13 | 1241 |