Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016805.2 | NP_058085.2 | 1 | 314 | ||
GenBank mRNA | AF073992.1 | - | 1 | 257 | ||
AK029944.1 | - | 1 | 260 | |||
AK037517.1 | - | 1 | 256 | |||
AK089058.1 | - | 1 | 315 | |||
AK145499.1 | - | 1 | 317 | |||
AK145682.1 | - | 1 | 316 | |||
AK159084.1 | - | 1 | 283 | |||
AK159614.1 | - | 1 | 319 | |||
AK159956.1 | - | 1 | 256 | |||
AK162264.1 | - | 1 | 318 | |||
AK165844.1 | - | 1 | 314 | |||
AK167200.1 | - | 1 | 316 | |||
AK168657.1 | - | 1 | 256 | |||
BC018353.1 | - | 1 | 159 | 3584692 |