Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018884.2 | NP_061372.2 | 3, 4 | 925 | ||
GenBank mRNA | AF127084.1 | - | 3, 4 | 925 | ||
AF127085.1 | - | 4, 5 | 769 | |||
AK143226.1 | - | 3, 4 | 439 | |||
AK173098.1 | - | 3, 4 | 884 | |||
HM560981.1 | - | 3, 4 | 916 |