Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021336.4 | NP_067311.4 | 6 | 799 | ||
GenBank mRNA | AK088438.1 | - | 6 | 574 | ||
AK151800.1 | - | 6 | 592 | |||
AK152546.1 | - | 6 | 592 | |||
AK153502.1 | - | 6 | 592 | |||
AK206837.1 | - | 5 | 362 | |||
BC013777.1 | - | 6 | 566 | 2651704 | ||
BC029642.1 | - | 6 | 566 | 3588138 |