Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021529.3 | NP_067504.2 | 3 | 368 | ||
GenBank mRNA | AB041577.1 | - | 3 | 357 | ||
AJ238247.2 | - | 2 | 195 | |||
AK013403.1 | - | 2 | 164 | |||
AK017550.1 | - | 2 | 151 | |||
AK028063.1 | - | 3 | 333 | |||
AK028254.1 | - | 2 | 287 | |||
AK033405.1 | - | 3 | 366 | |||
AK040589.1 | - | 2 | 196 | |||
AK087572.1 | - | 2 | 191 | |||
AK088240.1 | - | 2 | 277 | |||
AK163192.1 | - | 367 | ||||
BC024754.1 | - | 3 | 331 | 5375405 | ||
BC138295.1 | - | 2 | 164 | 8861314 | ||
BC138296.1 | - | 2 | 164 | 8861315 |