Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025375.2 | NP_001020546.1 | 4, 5 | 363 | ||
NM_001025376.2 | NP_001020547.1 | 4, 5 | 252 | |||
NM_023191.3 | NP_075680.1 | 4, 5 | 257 | |||
GenBank mRNA | AF309554.1 | - | 4, 5 | 215 | ||
AK010283.1 | - | 4, 5 | 203 | |||
AK012342.1 | - | 4, 5 | 269 | |||
AK013114.1 | - | 4, 5 | 221 | |||
AK049527.1 | - | 4, 5 | 221 | |||
AK077037.1 | - | 297 | ||||
AK142498.1 | - | 4, 5 | 220 | |||
AK153862.1 | - | 4, 5 | 290 | |||
AK167706.1 | - | 4, 5 | 290 | |||
BC023026.1 | - | 4, 5 | 202 | 5374890 | ||
BY081994.1 | - | 4, 5 | 309 | |||
BY165863.1 | - | 4, 5 | 223 | |||
EF011623.1 | - | 4, 5 | 200 |