Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025464.2 | NP_079740.1 | 4 | 362 | ||
GenBank mRNA | AK003010.1 | - | 4 | 359 | ||
AK007575.1 | - | 4 | 346 | |||
AK008902.1 | - | 4 | 333 | |||
BC034160.1 | - | 4 | 362 | 5049687 |